Learn More
Vortioxetine hydrobromide, MedChemExpress
Product Code. 16466498
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. Quantity unitSize
16466498 10 mg 10mg
16452597 50 mg 50mg
2 options
Product Code. 16466498
Brand: Medchem Express HY15414A
MedChemExpress Vortioxetine hydrobromide is a multimodal serotonergic agent, inhibits 5-HT1A, 5-HT1B, 5-HT3A, 5-HT7 receptor and SERT with Ki values of 15 nM, 33 nM, 3.7 nM, 19 nM and 1.6 nM, respectively.
Vortioxetine hydrobromide is a multimodal serotonergic agent, inhibits 5-HT1A, 5-HT1B, 5-HT3A, 5-HT7 receptor and SERT with Ki values of 15 nM, 33 nM, 3.7 nM, 19 nM and 1.6 nM, respectively.
Specifications
Chemical Name or Material Vortioxetine hydrobromide
Physical Form Solid
CAS 960203-27-4
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H23BrN2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym Lu AA21004 hydrobromide
Solubility Information DMSO : 50 mg/mL (131.80 mM; Need ultrasonic)
SMILES CC1=CC=C(SC2=CC=CC=C2N3CCNCC3)C(C)=C1.[H]Br
Molecular Weight (g/mol) 379.36
Formula Weight 379.36
Percent Purity 99.45%
Grade Research