Learn More
Vortioxetine.HBr, Lipomed
Description
Vortioxetine.HBr
Specifications
Specifications
| Chemical Name or Material | Vortioxetine Hydrobromide |
| CAS | 960203-27-4 |
| Analyte or Component Names | Vortioxetine Hydrobromide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H22 N2 S . Br H |
| Synonym | Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, hydrobromide (1:1), 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine hydrobromide, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine hydrobromide, Lu AA 21004 hydrobromide, Vortioxetine hydrobromide |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Br.Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1 |
| IUPAC Name | 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide |
| Industry Type | Pharmaceutical |
| Show More |