Learn More
Vortioxetine.HBr, Lipomed
Product Code. 30458095
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
30458095 100 mg 100mg
1 options
Product Code. 30458095
Brand: Lipomed LPMVOX1884HB100
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Vortioxetine.HBr
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Vortioxetine Hydrobromide
CAS 960203-27-4
Analyte or Component Names Vortioxetine Hydrobromide
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 S . Br H
Synonym Piperazine, 1-[2-[(2,4-dimethylphenyl)thio]phenyl]-, hydrobromide (1:1), 1-[2-[(2,4-Dimethylphenyl)thio]phenyl]piperazine hydrobromide, 1-[2-(2,4-Dimethylphenylsulfanyl)phenyl]piperazine hydrobromide, Lu AA 21004 hydrobromide, Vortioxetine hydrobromide
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Br.Cc1ccc(Sc2ccccc2N3CCNCC3)c(C)c1
IUPAC Name 1-[2-(2,4-dimethylphenyl)sulfanylphenyl]piperazine;hydrobromide
Industry Type Pharmaceutical
Molecular Weight (g/mol) 379.358
Quantity 100 mg
Formula Weight 378.077 g/mol