Learn More
Cole-Parmer™ Vortex-Genie™ 2 Shaker
Product Code. 11830170 Shop All Cole-Parmer Products
Change view
Click to view available options
Voltage:
220 V
Unit Size:
Each
Product Code. Voltage unitSize
11830170 220 V Each
1 options
Product Code. 11830170
Brand: Cole-Parmer™ 0472405
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Adapt your shaker to a variety of applications. Cole-Parmer™ Vortex-Genie™ 2 Shaker features variable speed shaking which allows you to adjust the level of mixing from gentle shaking to vigorous vortexing.
Shaker allows you to control speed intensity by varying pressure to the head. Use the pop-off cup for single tube shaking or the 3 in. platform for multiple tube or flask shaking. Enhanced vortex action for maximum high-speed touch mixing. Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop. Includes pop-off cup and 3 in. platform. Power cord and plug (Euro plug on 230 VAC model).
Specifications
Speed Range 600 to 2700 rpm
Electrical Requirements 220 V, 50 Hz
Voltage 220 V
Product Type Shaker
Frequency 50 Hz
Height (Metric) 152.4 mm
Width (Metric) 123.8 mm