missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VorTemp™ 56 Shaking Incubator and Temperature Validation Kit

Product Code. p-9053045
Change view
Click to view available options
Voltage:
230 V
Unit Size:
Each
Product Code. Voltage unitSize
16329533 230 V Each
1 options
This item is not returnable. View return policy

Product Code. 16329533

Brand: Invitrogen™ QP0704

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.

The VorTemp 56 microplate shaking incubator is designed for simultaneous heating and mixing of QuantiGene Plex assays. The temperature is adjustable over a broad range, and mechanical convection provides a uniform and stable environment within the chamber.

VorTemp features:

  • A microprocessor to control all parameters including temperature and timed operation
  • Self-diagnostic software to continuously monitor for proper temperature and shaking speed, alerting the user to any errors
  • Heating and shaking that are controlled by feedback loops which precisely maintain chosen parameters, independent of load or voltage fluctuations
  • Sturdy construction for stability during high speed shaking
  • An insulated chamber to maintain temperature and reduce operating noise
TRUSTED_SUSTAINABILITY

Specifications

Voltage 230 V
Temperature Range (Metric) 4°C to 99.5°C
Product Type Shaking Incubator
Material Coated Steel
Humidity Range 85%
Includes 1 VorTemp 56 Shaking Incubator, 1 QuantiGene Temperature Validation Kit
Quantity 1 Ea.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.