Learn More
Socorex™ Acura™ 8-channel Micropipette Volumetric Module
Click to view available options
Volume (Metric):
10 μL
50 μL
350 μL
Unit Size:
Each
Description
- One of 27 available volumetric assemblies.
- Increase versatility and save costs.
- Disassembles and reassembles without tools.
- Calibration specificities of extra modules - as set by factory QC or by pipette owner - are retained in instrument memory.
Specifications
Specifications
| For Use With | 8 Channel Pipette |
| Number of Channels | 8 |
| Product Line | Acura |
| Type | Micropipette Volumetric Module |
| Volume (Metric) | 350 μL |
| For Use With (Application) | 8 Channel Pipette |
| For Use With (Equipment) | Acura™ Electro 926 Micropipette |
| Quantity | 1 Each |