Learn More
KARTELL Volumetric Flask
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
Unit Size:
Each
Description
- Volumetric flask made of transparent fluorinated resin with high chemical and temperature resistance (-250°C to +270°C).
- Supplied with standard DIN stoppers in PFA.
- Individually calibrated ringmark according to DIN EN ISO 1042, class “A”, Standards.
- Laser engraved lot number.
Specifications
Specifications
| Capacity (Metric) | 250 mL |
| Material | PFA |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Graduated | No |
| Mouth Style | Narrow |
| Sterility | Non-sterile |
| Bottom Shape | Flat |
| Surface Treatment | Non-treated |