Learn More
BRAND™ Volumetric Flask
Product Code. 15941322 Shop All Brand Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Each
Product Code. 15941322
Brand: BRAND™ 937262
Only null left
Add to basket
Add to basket
With glass stopper
  • Boro 3.3
  • DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • TRUSTED_SUSTAINABILITY
    Specifications
    Closure Material Glass
    Capacity (Metric) 250 mL
    Material Borosilicate Glass (3.3)
    Class A