Learn More
Poulten Graf™ Volac™ USP Class A Soda Lime Glass Bulb Pipette with Works Certificate
Description
- Individually numbered and meets the accuracies of USP/ISO/BS and DIN standards
- Amber stain graduation
- Calibrated for delivery
Specifications
Specifications
| Certifications/Compliance | Certificate of calibration/Satisfying USP/ISO/BS and DIN accuracies |
| Class | A |
| Material | Soda Lime Glass |
| Graduated | Yes |
| Quantity | 5 Pack |
| Tolerance | 0.03 mL |
| Volume (Metric) | 25 mL |
| Type | Bulb Pipet |