Learn More
Poulten Graf™ Volac™ Borosilicate Glass Burettes with Glass Key
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Graduations:
0.02 mL
0.05 mL
0.1 mL
Unit Size:
Each
Description
- Removable glass jets are inscribed with instrument number for traceability
- Flow time inscribed on the instrument
- Certificate specifying accuracy at three points provided
- Produced according to BS ISO 385
Specifications
Specifications
| Color | Blue |
| Capacity (Metric) | 25 mL |
| Length (Metric) | 820 mm |
| Material | Borosilicate Glass |
| Graduations | 0.05 mL |
| Includes | Interchangeable Glass Key in Glass Barrel |
| Stopcock | Straight |
| Stopcock Material | Glass |
| Tolerance | ±0.03 mL |
| Type | Volumetric Buret |
| Show More |