Learn More
Sartorius™ Vivacon™ Polycarbonate/Polypropylene Centrifugal Unit
Shop All Sartorius Products
Click to view available options
MWCO:
100 kDa
30 kDa
Unit Size:
Pack of 25
Specifications
Specifications
| For Use With (Equipment) | PCR |
| Material | Polycarbonate, Polypropylene |
| MWCO | 100 kDa |
| For Use With (Application) | Concentration, Desalting, buffer exchange |
| Housing Material | Polycarbonate, Polypropylene |
| Quantity | 25/Pk. |
| Sample Type | DNA |
| Type | Vivacon 500 Centrifugal Unit |
| Volume (Metric) | 0.1 to 0.5 mL |