Learn More
Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint
Product Code. 11897900
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methylene chloride
Unit Size:
Each
Pack of 12
Pack of 6
Product Code. 11897900
Brand: Vitlab™ 1332879
Only null left
Add to basket
Add to basket
Identify media quickly and easily with these color-coded wash bottles. VITsafe™ Narrow-Mouth Wash Bottles with Imprint feature a precise spray jet and optimized medium backflow through the continuous and finely drawn tip of the spray tube.
  • Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
  • VENT-CAP practically eliminates leakage and prevents almost all static overpressure
  • Color-coded for easy visual identification of the media
Bottles feature durable safety imprint in accordance with Directive (EC) No. 1272/2008 (GHS), as well as with all important information:
  • Material name in German, English, French and Spanish
  • Chemical formula, CAS No., hazard pictogram, signal word
  • Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Specifications
Capacity (Metric) 500 mL
For Use With (Application) Saving and storing
Includes Spray tube and imprint
Material LDPE
No. per Pack 6 Pack
Closure Color Red
Closure Included Yes
Mouth Narrow
Height (Metric) 180 mm
Product Type Wash Bottle
Chemical Label Methylene chloride
Closure Material Polypropylene
Diameter (Metric) 74 mm
Closure Type Vented