Learn More
Vitlab™ VITsafe™ Narrow-Mouth Wash Bottles with Imprint
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Chemical Label:
Acetone
Distilled Water
Ethanol
Ethyl Acetate
Isopropanol
Methylene chloride
Tamaño de la unidad:
Each
Pack of 12
Pack of 6
Descripción
- Bottles made from LDPE (or PP for Acetone and MEK), with polypropylene spray tube
- VENT-CAP practically eliminates leakage and prevents almost all static overpressure
- Color-coded for easy visual identification of the media
- Material name in German, English, French and Spanish
- Chemical formula, CAS No., hazard pictogram, signal word
- Risk phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
Especificaciones
Especificaciones
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Saving and storing |
| Includes | Spray tube and imprint |
| Material | LDPE |
| No. per Pack | 12 Pack |
| Closure Color | Red |
| Closure Included | Yes |
| Mouth | Narrow |
| Height (Metric) | 180 mm |
| Product Type | Wash Bottle |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido