Learn More
Ismatec™ Viton™ Microbore 2-Stop Tube Sets
Product Code. p-9049025
Click to view available options
Diameter (Metric) Inner:
0.89 mm
1.42 mm
2.79 mm
Flow Rate:
130 mL/min
200 mL/min
30 mL/min
67 mL/min
Unit Size:
Pack of 12
Product Code. 11719829
Brand: Ismatec™ 9642826
Only null left
Add to basket
Add to basket
Choose the size and formulation to suit your flow rate and application needs.
  • Genuine Ismatec™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
Specifications
Certifications/Compliance FDA approved; Prop 65 NA (no Prop 65 chemicals)
Diameter (Metric) Inner 0.89 mm
Material Viton
Overall Length (English) 1.25 ft.
Overall Length (Metric) 0.38 m
For Use With Pump Series L/S
Min. Operating Temperature (Metric) 0°C
Packaging Quantity 12 / Pk.
Flow Rate 30 mL/min
Maximum Pressure 25.3 psi, 1.75 bar
Max. Operating Temperature (Metric) 150°C
Product Type Stopped Pump Tubing