Learn More
BRANDTECH™ VITLAB™ Polypropylene Graduated Cylinders with Blue Scale, Class B
Product Code. p-10054059
Click to view available options
Unit Size:
Pack of 3
Product Code. 10629682
Brand: BRANDTECH™ VITLAB™ 653081
Only null left
Add to basket
Add to basket
Polypropylene Graduated Cylinders are highly transparent with easy-to-read, embossed blue graduations.
  • Highly Transparent: Ensures clarity for easy content viewing
  • Molded Blue Gradations and Ring Marks: Easily readable markings at primary scale
  • Calibrated ‘To Contain’: Compliant with Class B tolerances
  • Hexagonal Base with Bottom Studs: Provides high stability and prevents tipping
  • Thermal Stress Resistant: Can withstand temperatures up to 80°C without exceeding tolerance limits, but to maintain markings, avoid cleaning at temperatures above 60°C
Specifications
Material Polypropylene
Tolerance ±20 mL
Certifications/Compliance DIN 12681, ISO 6706
Class Class B
Calibration To Contain
Disposable Reusable
Diameter (English) 3.81 in.
Scale Blue molded graduations
Diameter (Metric) 97 mm
Graduations 20 mL
Autoclavable Autoclavable
Max. Temperature 60°C
Subdivision 20mL
Shape Round
Sterility Non-sterile
Base Type Hexagonal
Height (English) 18.97 in.
Height (Metric) 482 mm
Type Graduated Cylinder