missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vitamin D binding protein Mouse anti-Human, Clone: 1E4D10, Proteintech
Shop All Proteintech ProductsDescription
GC (vitamin D-binding protein) is involved in vitamin D transport/storage, scavenging of extracellular G-actin, enhancement of the chemotactic activity of C5 alpha for neutrophils in inflammation and macrophage activation.
Specifications
Specifications
| Antigen | Vitamin D binding protein |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 1E4D10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | GC |
| Gene Accession No. | P02774 |
| Gene Alias | DBP, DBP/GC, GC, Gc globulin, VDBP,GC, Vitamin D binding protein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?