Learn More
Vitamin A palmitate, MP Biomedicals™
Product Code. 11497481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497481 100 g 100g
11477481 5 g 5g
11487481 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11497481 Supplier MP Biomedicals Supplier No. 0210326380
  • ONLINE EXCLUSIVE PRICE
    3890.00 NOK
    3703.16 NOK / 100g
    Save 186.84 NOK
    4% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Chemical Identifiers
CAS 79-81-2
Molecular Formula C36H60O2
Molecular Weight (g/mol) 524.874
InChI Key VYGQUTWHTHXGQB-FFHKNEKCSA-N
Synonym retinyl palmitate, vitamin a palmitate, retinol palmitate, retinol, hexadecanoate, arovit, all-trans-retinyl palmitate, optovit-a, retinyl hexadecanoate, aquapalm, dispatabs tabs
PubChem CID 5280531
ChEBI CHEBI:17616
IUPAC Name [(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC=C(C)C=CC=C(C)C=CC1=C(CCCC1(C)C)C
Specifications
Color Yellow to Orange
Quantity 100 g
Formula Weight 524.9
Physical Form Powder
Chemical Name or Material Vitamin A palmitate