Learn More
R&D Systems™ VisUCyte Antigen Retrieval Reagent-Basic
Product Code. 18772388
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18772388 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18772388 Supplier R&D Systems™ Supplier No. VCTS021
  • ONLINE EXCLUSIVE PRICE
    1644.10 NOK
    1461.60 NOK / 50mL
    Save 182.50 NOK
    11% Off
 Restricted Item
Estimated Shipment: 17-11-2026
VisUCyte™ Antigen Retrieval Reagents are three different antigen retrieval systems that utilize heat-induced recovery of cell and tissue antigens.
  • Unlike Universal solution, Acidic and Basic solutions may dramatically enhance immunoreactivity but may also affect tissue morphology.
  • The optimal antigen unmasking condition is a function of temperature (90-100°C) and incubation time (up to 30 minutes) and should be established by each individual investigator.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at room temperature. Do not use past expiration date.
Product Line R&D Systems
Quantity 50 mL