Learn More
VistaLab™ Technologies Seal Kits for MLA™ Brand Pipets
Provide materials for maintenance of MLA D-Tipper™, Precision, and two-stroke pipetters
Brand: VistaLab™ Technologies 9035
Additional Details : Weight : 0.17240kg
Description
Each kit contains seals and instructions
Order Info
Lubricant sold separately (P/N 22-146-318)
Specifications
351 to 1000 μL | |
Lubricant, seals and instructions | |
2 in. | |
1 in. |
351 to 1000 μL MLA™ D-Tipper™ and Precision Pipette | |
1 Ea. | |
1 in. |