Learn More
VistaLab™ Technologies Seal Kits for MLA™ Brand Pipets
Provide materials for maintenance of MLA D-Tipper™, Precision, and two-stroke pipetters
Brand:  VistaLab™ Technologies 9032
Additional Details : Weight : 0.17240kg
 View more versions of this product
Product Code. 16415134
  • 232.80 NOK / 1 set
Estimated Shipment: 16-05-2024
Description
Each kit contains seals and instructions
Order Info
Lubricant sold separately (P/N 22-146-318)
Specifications
Specifications
31 to 120 μL
Lubricant, seals and instructions
2 in.
1 in.
31 to 120 μL MLA™ D-Tipper™ and Precision Pipette
1 Ea.
1 in.