Learn More
VistaLab™ Technologies MLA™ Brand Digital Pipetters
Click to view available options
Volume (Metric):
10 to 50 μL
200 to 1000 μL
50 to 200 μL
Accuracy:
±1%
±2%
Color:
Gray
Green
Red
Compatible Tips:
0.2 mL Tip
1mL Tip
Unit Size:
Each
Description
Three adjustable models cover volume range: 10 to 1000μL. To adjust volume, simply turn volume selector knob near top of pipetter. Four-digit reading is locked in by click stops, ensuring accuracy.
- One-handed operation
- D-Tipper™ feature allows you to pick up and dispose of tips without touching them
- Volume range conveniently marked on all models
- Factory calibrated for accurate delivery
- Easy in-lab recalibration
- Made of lightweight aluminum and stainless steel to assure durability and long life
Order Info
See MLA Brand tips Mfr. Nos. 2025, 2026, 2027, 2028, 4025, 4026, 9025, 9026, 9029
Specifications
Specifications
| Accuracy | ±1% |
| Compatible Tips | 0.2 mL Tip |
| Autoclavable | Not Autoclavable |
| Color | Red |
| Disposable | Reusable |
| For Use With | 0.2 mL Tip |
| Grip Type | Ergonomic |
| Material | Aluminum, Stainless Steel |
| Number of Channels | 1 |
| Product Line | MLA Brand |
| Show More |