Learn More
VistaLab™ Technologies MLA™ Brand Digital Pipetters
GREENER_CHOICE
Product Code. 11985852
Change view
Click to view available options
Volume (Metric):
10 to 50 μL
200 to 1000 μL
50 to 200 μL
Accuracy:
±1%
±2%
Color:
Gray
Green
Red
Compatible Tips:
0.2 mL Tip
1mL Tip
Unit Size:
Each
Product Code. Volume (Metric) Accuracy Color Compatible Tips unitSize
11985852 10 to 50 μL ±2% Green 0.2 mL Tip Each
11905862 200 to 1000 μL ±1% Gray 1mL Tip Each
11995852 50 to 200 μL ±1% Red 0.2 mL Tip Each
3 options
Product Code. 11985852
Brand: VistaLab™ Technologies 1136
Only null left
Add to basket
Add to basket
Perfect for tests that require multiple or nonstandard volumes
Three adjustable models cover volume range: 10 to 1000μL. To adjust volume, simply turn volume selector knob near top of pipetter. Four-digit reading is locked in by click stops, ensuring accuracy.
  • One-handed operation
  • D-Tipper™ feature allows you to pick up and dispose of tips without touching them
  • Volume range conveniently marked on all models
  • Factory calibrated for accurate delivery
  • Easy in-lab recalibration
  • Made of lightweight aluminum and stainless steel to assure durability and long life
Order Info
See MLA Brand tips Mfr. Nos. 2025, 2026, 2027, 2028, 4025, 4026, 9025, 9026, 9029
Specifications
Accuracy ±2%
Compatible Tips 0.2 mL Tip
Autoclavable Not Autoclavable
Color Green
Disposable Reusable
For Use With 0.2 mL Tip
Grip Type Ergonomic
Material Aluminum, Stainless Steel
Number of Channels 1
Product Line MLA Brand
Programmability No
Sterility Non-sterile
Type Digital Pipetter
Volume (Metric) 10 to 50 μL
For Use With (Application) Multiple or Non-Standard Volumes
For Use With (Equipment) 0.2 mL Tip
Includes Nozzle Insert, Calibration Key
Quantity 1 Ea.
Warranty Limited Lifetime
Length (English) 9 in.
Width (English) 2 in.
Height (English) 1.5 in.