Learn More
VistaLab™ Technologies Cap Opener
Specifications
Specifications
| Compatible Storage Tubes | Tubes with 8 mm Caps |
| Length (English) | 3 in. |
| No. per Pack | 1 Ea. |
| Operation | Manual |
| Width (English) | 3 in. |
| Type | Cap Opener Kit |
| For Use With (Equipment) | Ovation electronic single channel and adjustable and fixed volume models |
| Size | 11 mm |
| For Use With (Application) | Ideal easy-to-use ergonomic solution for opening PCR or micro tubes while pipetting |