Learn More
Visomitin, MedChemExpress
Product Code. 16472586
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16472586 25 mg 25mg
16453038 10 mg 10mg
16429606 50 mg 50mg
16425777 5 mg 5mg
16404346 100 mg 100mg
5 options
Product Code. 16472586
Brand: Medchem Express HY100474
MedChemExpress Visomitin (SKQ1) is a mitochondrial-targeted antioxidant with the high mitochondrion membrane penetrating ability and potent antioxidant capability.
Visomitin (SKQ1) is a mitochondrial-targeted antioxidant with the high mitochondrion membrane penetrating ability and potent antioxidant capability.
Specifications
Chemical Name or Material Visomitin
Physical Form Solid
CAS 934826-68-3
Color Brownish Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C36H42BrO2P
For Use With (Application) COVID-19-immunoregulation
Synonym SKQ1
Solubility Information DMSO : 100 mg/mL (161.92 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (80.96 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (5.39 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C(CCCCCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)=C4)C(C)=C(C)C4=O.[Br-]
Molecular Weight (g/mol) 617.6
Formula Weight 617.6
Percent Purity 95.57%
Grade Research