Learn More
Macherey-Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits
Run a wide range of water testing assays. Macherey Nagel™ Visocolor™ Eco Colorimetric and Titrimetric Test Kits avoid hazardous substances wherever possible.
Brand:  Macherey-Nagel™ 935042
Additional Details : Weight : 0.06300kg Transport : UN number : 3316 Chem class : 9 Pack group : II
 View more versions of this product
Product Code. 10675391
  • 286.00 NOK / 1mL
Estimated Shipment: 22-05-2024
Description
  • Colorimetric and titrimetric test kits
  • Avoids hazardous substances wherever possible
Specifications
Specifications
To determine the total hardness of water
1 drop = 1 °d (corresponds to 18 mg/l CaCO3)
Complexometric titration
30mL Special chemical reagent
100 Tests with an average hardness of 10 °d
Visocolor Alpha Hardness (Total)
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOR alpha total Hardness
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : 2-propanol