Learn More
3M™ Visionair™ Peel-Off Visor Cover
Description
- Peel-off lens cover for Visionair system
Specifications
Specifications
| Color | Transparent |
| For Use With (Equipment) | Visionair systems |
| Product Type | Visor Cover |