missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
visinin-like 1 Polyclonal antibody specifically detects visinin-like 1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | visinin-like 1 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HLP3Hippocalcin-like protein 3, HPCAL3, HUVISL1, VILIP-1, VILIPVLP-1, visinin-like 1, visinin-like protein 1, VISL1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 2-191 of human visinin-like 1 (NP_003376.2).,, Sequence:, MGKQNSKLAPEVMEDLVKSTEFNEHELKQWYKGFLKDCPSGRLNLEEFQQLYVKFFPYGDASKFAQHAFRTFDKNGDGTIDFREFICALSITSRGSFEQKLNWAFNMYDLDGDGKITRVEMLEIIEAIYKMVGTVIMMKMNEDGLTPEQRVDKIFSKMDKNKDDQITLDEFKEAAKSDPSIVLLLQCDIQK |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?