Learn More
Poulten Selfe™ Viscometer Holder
Click to view available options
Product Type:
U-tube Viscometer Holder
Ubbelohde Viscometer Holder
Size:
1 to 4
G, H
Unit Size:
Each
Specifications
Specifications
| Product Type | Ubbelohde Viscometer Holder |
| For Use With (Equipment) | Ubbelohde Suspended Level Viscometer |
| For Use With (Application) | 50 mm Bath Opening |
| Size | 1 to 4 |