Learn More
SI Analytics™ ViscoClock Plus
Product Code. 15679990 Shop All SI Analytics Products
Click to view available options
For Use With (Equipment):
Ubbelohde Viscometer
Unit Size:
Each
Product Code. 15679990
Brand: SI Analytics™ 285417900
Only null left
Add to basket
Add to basket
ViscoClock plus is an electronic timing unit for Ubbelohde type glass capillary viscometers used to determine kinematic and relative viscosity.
  • Specially designed for Ubbelohde viscometers
  • Allows automatic measurement of flow times
  • Stand in high performance engineering PPA for temperatures up to 150°C
  • Convenient handling of Ubbelohde viscometer due to electromagnetic venting valve
  • Data storage including time, date, viscometer and sample identification number
TRUSTED_SUSTAINABILITY
Specifications
Product Type Electronic Timing Unit
For Use With (Equipment) Ubbelohde Viscometer