Learn More
3M™ Virtua™ Safety Glasses
Product Code. 17981030
Change view
Click to view available options
Frame Type:
Frameless
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Frame Type unitSize
17981030 Frameless Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17981030 Supplier 3M™ Supplier No. 7100244069
  • ONLINE EXCLUSIVE PRICE
    1400.00 NOK
    1002.08 NOK / Pack of 20
    Save 397.92 NOK
    28% Off
View all promotions
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
Virtua™ Safety Glasses are rimless safety glasses with a wraparound design for full coverage and contoured flexible temples to ensure comfort. The integrated side-shields provide added protection from flying objects and the lens features an anti-scratch coating for increased durability.
  • Economical basic unisex safety glasses for single use protection
  • Wraparound design improves the security of fit and prevents slipping
  • Design provides maximum clarity, excellent coverage and a wide field of vision
  • Easy to clean
  • Economical bulk package available (5-pack)
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
Material Plastic
Disposable Reusable
Lens Material Plastic
Frame Material Plastic
UV Protection None
Frame Type Frameless