Learn More
Day Impex™ Virkon™ Virucidal Disinfectant
Description
- Use biocides safely. Always read the label and product information before use
- Widest proven spectrum of any disinfectant - proven effective in independent tests against all 18 virus families affecting man and animals, including AIDS (HIV) and Hepatitis B
- Excellent bactericide and fungicide - cleans and disinfects work surfaces and equipment in a single, one-step operation
- Can be applied in powder form direct to spillages of blood and other body fluids
- Easily dissolves to make a liquid disinfectant
- Odourless, non-irritant to skin and eyes, non-toxic vapour phase, exceptionally low oral toxicity
- Under COSHH requirements, no maximum exposure limit is required
- Non-corrosive, non-bleaching and biodegradable
- Any viral disease transmitted via surfaces or instruments contaminated with infected blood, body fluids, faeces or saliva can be controlled by correct application
- For cleaning and disinfecting work surfaces and equipment
Specifications
Specifications
| Product Type | Virucidal Disinfectant |
| Container Type | Squeeze Pack |
| For Use With (Application) | Virucidal Disinfectant |
| Quantity | 6/Pk. |
| Volume (Metric) | 500 g |
Safety and Handling
Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H336-May cause drowsiness or dizziness.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.