missing translation for 'onlineSavingsMsg'
Learn More

Vipivotide tetraxetan Linker, MedChemExpress

Product Code. p-200099894
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
This item is not returnable. View return policy

Product Code. 16499865

Brand: Medchem Express HY43869

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Vipivotide tetraxetan Linker (PSMA-617 Linker) is a nonclaevable peptide linker for synthesis of Vipivotide tetraxetan (PSMA-617).

Vipivotide tetraxetan Linker (PSMA-617 Linker) is a nonclaevable peptide linker for synthesis of Vipivotide tetraxetan (PSMA-617).

Specifications

Chemical Name or Material Vipivotide tetraxetan Linker
CAS 1703768-74-4
Color White
Purity Grade Notes Research
Molecular Formula C33H45N5O9
Quantity 250 mg
Synonym PSMA-617 Linker
Solubility Information DMSO : 270 mg/mL (411.75 mM; Need ultrasonic)
SMILES O=C(O)[C@H](CCCCNC([C@H](CC1=CC=C2C=CC=CC2=C1)NC([C@H]3CC[C@H](CN)CC3)=O)=O)NC(N[C@H](C(O)=O)CCC(O)=O)=O
Molecular Weight (g/mol) 655.74
Formula Weight 655.74
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.