missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Violet Red Bile Glucose Agar (Contact)

Product Code. 15812467
Change view
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Product Code. Quantity unitSize
15812467 10 Plates Pack of 10
1 options
This item is not returnable. View return policy

Product Code. 15812467

Brand: Thermo Scientific™ PO5323C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize microbial isolation with this agar, providing targeted detection of glucose-fermenting bacteria.

Optimize microbial isolation with Violet Red Bile Glucose Agar (Contact), providing targeted detection of glucose-fermenting bacteria.

TRUSTED_SUSTAINABILITY

Specifications

Color Transparent
Content And Storage 2°C to 12°C
Format 90 mm
Manufacturing Origin Germany
Product Type Agar Plate
Fill Volume 17 mL
Quantity 10 Plates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.