Learn More
Vinylene Carbonate (Contains Up to 2% BHT), TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Vinylene Carbonate |
| CAS | 872-36-6 |
| Molecular Formula | C3H2O3 |
| Quantity | 25 g |
| Synonym | 1,3-Dioxol-2-one, Cyclic Vinylene Ester Carbonic Acid, LBG 84922, NSC 20980, NSC 47201, Vinylidene Carbonate |
| InChI Formula | InChI=1S/C3H2O3/c4-3-5-1-2-6-3/h1-2H |
| SMILES | O=C1OC=CO1 |
| IUPAC Name | 1,3-dioxol-2-one |
| Molecular Weight (g/mol) | 86.05 |
| Formula Weight | 86.0004 |