Learn More
Masterflex™ Vinyl Tubing
Product Code. 15218314
Click to view available options
Diameter (Metric) Inner:
1.6 mm
Unit Size:
15m
Product Code. 15218314
Brand: Masterflex™ 0640501
Only null left
Add to basket
Add to basket
General-purpose laboratory tubing with fair chemical and abrasion resistance.
  • Applications: General purpose laboratory use.
  • Fair chemical and abrasion resistance.
  • Odorless.
  • Nontoxic.
  • Transparent.
  • Manufactured according to GMP.
Specifications
Certifications/Compliance 3A;European Pharmacopoeia;NSF-51;ADCF;FDA;Non-Phthalate;USP;Non-DEHP;NSF-61
Autoclavable Not Autoclavable
Diameter (Metric) Inner 1.6 mm
Material Vinyl
Diameter (Metric) Outer 3.175 mm
Overall Length (Metric) 15.2 m
Thickness (Metric) 0.8 mm
Min. Operating Temperature (Metric) -34°C
Sterilization Method ETO
Color Clear
Maximum Pressure 40 psi
Max. Operating Temperature (Metric) 74°C