Learn More
Vinyl methacrylate, 98%, stab. with 200ppm 4-methoxyphenol, Thermo Scientific™
Catalog Number 11382147
Click to view available options
Quantity:
5 g
25 g
4245-37-8
C6H8O2
112.128
MFCD00059329
FFYWKOUKJFCBAM-UHFFFAOYSA-N
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11
77921
ethenyl 2-methylprop-2-enoate
CC(=C)C(=O)OC=C
4245-37-8
C6H8O2
112.128
MFCD00059329
FFYWKOUKJFCBAM-UHFFFAOYSA-N
vinyl methacrylate, methacrylic acid, vinyl ester, methacrylic acid vinyl ester, 2-methyl-2-propenoic acid, ethenyl ester, 2-propenoic acid, 2-methyl-, ethenyl ester, methacrylic acid, vinyl ester 8ci, vinylmethacrylate, vinyl 2-methylacrylate #, acmc-1cr11
77921
ethenyl 2-methylprop-2-enoate
CC(=C)C(=O)OC=C
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Density 0.94
Boiling Point 111°C to 112°C
Flash Point 16°C (60°F)
Odor Pungent
Refractive Index 1.435
Quantity 5 g
UN Number UN3272
Beilstein 1743336
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material Vinyl methacrylate, stabilized with 200ppm 4-methoxyphenol