Learn More
Vinyl Acetate, Sigma-Aldrich™
Product Code. 15826350
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15826350 100 mL 100mL
15818940 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15826350 Supplier Sigma Aldrich Supplier No. 8031840100
 Restricted Item
Estimated Shipment: 19-11-2026
Stabilized for synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 108-05-4
Specifications
Melting Point -93°C
Density 0.93 g/cm3 at 20°C
Boiling Point 71°C to 73°C/1013 hPa
MDL Number MFCD00008713
Quantity 100 mL
Synonym Vinyl acetate, Acetic acid vinyl ester, Vinyl acetate
Molecular Weight (g/mol) 86.09
Grade Research