missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vinculin, Mouse anti-Human,Mouse, Clone: VCL/2573, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79807-100UG
This item is not returnable.
View return policy
Description
Vinculin Monoclonal specifically detects Vinculin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Vinculin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P18206, P18206 | |
| VCL | |
| Recombinant fragment (around aa 174-322) of human Vinculin protein (exact sequence is proprietary) | |
| 100 μg | |
| Cell Biology, Cellular Markers, Phospho Specific | |
| 7414 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| VCL/2573 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction