Learn More
Vileda Professional™ Telescopic Handle, short
Description
- Constructed of aluminum
- Adjustable length of 50 - 90cm
- Extends with twist of the wrist
- Anti-slip top grip to reduce slipping on walls
- Ergonomic grip allows blood circulation in hands and fingers, thereby reducing fatigue
- Resists autoclave, gamma and ETO processes
- Fits CE Squeegee and other commonly used frames
Specifications
Specifications
| Product Type | Telescopic Handle, Short |
| Color | Blue, Silver |
| For Use With (Equipment) | Swep Duo Plus Frame |
| For Use With (Application) | Floors, Walls and Ceilings, ISO 5, LAR, Production, Labs |