Learn More
Vileda Professional™ CleanTech™ Duo Microfiber Laminated Mop, Sterile
Product Code. 17882496
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 45
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
17882496 Sterile Pack of 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17882496 Supplier Vileda Professional™ Supplier No. 147544
  • ONLINE EXCLUSIVE PRICE
    21125.00 NOK
    16631.15 NOK / Pack of 45
    Save 4493.85 NOK
    21% Off
Estimated Shipment: 16-12-2026
For cleaning/disinfecting floors, walls and ceilings. Vileda Professional CleanTech Duo Microfiber Laminated Mop, Sterile are designed for use in cleanrooms up to class ISO 5/GMP A and B/FED209D Class 100.
  • Microfiber laminated cleanroom foam
  • Double sided for effective cleaning and disinfecting
  • Adhesive-free lamination techniques used to prevent additional contamination to products
  • Suitable for bucket and pre-prepared method
  • Proved bacteria reduction of 99.99%
  • Validated to the 10-6 SAL per ISO 11137
  • Individually double bagged with Lot # and Indicator Dot on each package
  • Certificate of Sterility available upon request