Description
- Viewing window with triple insulating glass in door(s)
- 15 or 30 W interior lighting, depending on the model
Specifications
Specifications
| Description | Viewing window and interior lighting - 230 V option model, viewing window (triple insulating glass) in door, 240 x 240 mm, and 15 W interior |
| For Use With (Equipment) | BINDER Chambers ED 56 |
| Type | Viewing Window with Interior Lighting |
| Insulation | Triple Insulating Glass |
| Width (Metric) | 240 mm |
| Height (Metric) | 240 mm |
| Wattage | 15 W |