Learn More
Human alpha-Galactosidase A/GLA Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6146-SP
This item is not returnable.
View return policy
Description
alpha-Galactosidase A/GLA Polyclonal specifically detects alpha-Galactosidase A/GLA in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| alpha-Galactosidase A/GLA | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Agalsidase, agalsidase alfa, Agalsidase alpha, Alpha-D-galactosidase A, Alpha-D-galactoside galactohydrolase, alpha-D-galactoside galactohydrolase 1, alpha-gal A, alpha-galactosidase A, EC 3.2.1, EC 3.2.1.22, GALA, galactosidase, alpha, melibiase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2717 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P06280 | |
| GLA | |
| Chinese hamster ovary cell line CHO-derived recombinant human alpha-Galactosidase A/GLA Leu32-Leu429 Accession # P06280 | |
| 25 μg | |
| Primary | |
| Detects human alpha-Galactosidase A/GLA in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction