Learn More
PX-12, MedChemExpress
Product Code. 16486296
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16486296
Brand: Medchem Express HY13734
MedChemExpress PX-12(IV-2) is an irreversible inhibitor of Thioredoxin-1 (Trx-1); inhibits the growth of MCF-7 and HT-29 cells with IC50 values of 1.9 and 2.9 μM, respectively.
PX-12(IV-2) is an irreversible inhibitor of Thioredoxin-1 (Trx-1)
Specifications
Chemical Name or Material PX-12
Physical Form Solid
CAS 141400-58-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C7H12N2S2
For Use With (Application) Cancer-programmed cell death
Synonym IV-2
Solubility Information Ethanol : 50 mg/mL (265.52 mM; Need ultrasonic) ∣DMSO : ≥ 44.7 mg/mL (237.37 mM)
SMILES CCC(SSC1=NC=CN1)C
Molecular Weight (g/mol) 188.31
Formula Weight 188.31
Percent Purity 98.0%
Grade Research