Learn More
(Z)-9-Propenyladenine, MedChemExpress
Product Code. 16480499
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480499 1 mg 1mg
16482768 10 mg 10mg
16475507 5 mg 5mg
16464226 100 mg 100mg
16459466 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16480499 Supplier Medchem Express Supplier No. HY100079A
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress (Z)-9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase (NtART) inhibitor, which blocks reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV.
(Z)-9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase (NtART) inhibitor, which blocks reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV.
Specifications
Chemical Name or Material (Z)-9-Propenyladenine
CAS 1464851-21-5
Color White
Purity Grade Notes Research
Molecular Formula C8H9N5
Quantity 1 mg
Synonym (Z)-Mutagenic Impurity of Tenofovir Disoproxil
Solubility Information DMSO : ≥ 100 mg/mL (570.81 mM)
SMILES NC1=C2N=CN(/C=C\C)C2=NC=N1
Molecular Weight (g/mol) 175.19
Formula Weight 175.19
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus