Learn More
MedChemExpress pGlu-Pro-Arg-MNA monoacetate
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| CAS | 2070009-26-4 |
| Solubility | H2O : ≥ 25 mg/mL (42.19 mM) |
| Content And Storage | Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Quantity | 5 mg |
| Molecular Weight (g/mol) | 592.6 g/mol |
| Product Type | Biochemical Assay Reagents |
| Purity or Quality Grade | 99.0% |
| Shipping Condition | Room temperature |