Learn More
GSK3326595, MedChemExpress
Product Code. 16443418
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Product Code. 16443418
Brand: Medchem Express HY101563
MedChemExpress GSK3326595 (EPZ015938) is a potent, selective, reversible inhibitor of protein arginine methyltransferase 5 (PRMT5) with an IC50 of 6.2 nM.
GSK3326595 (EPZ015938) is a potent, selective, reversible inhibitor of protein arginine methyltransferase 5 (PRMT5) with an IC50 of 6.2 nM.
Specifications
Chemical Name or Material GSK3326595
CAS 1616392-22-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H32N6O3
Quantity 10 mg
Synonym EPZ015938
Solubility Information DMSO : 66.67 mg/mL (147.32 mM; Need ultrasonic)
SMILES O=C(C)N1CCC(NC2=NC=NC(C(NC[C@H](O)CN3CCC(C=CC=C4)=C4C3)=O)=C2)CC1
Molecular Weight (g/mol) 452.55
Formula Weight 452.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death