Learn More
Mavatrep, MedChemExpress
Product Code. 16433237
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16433237
Brand: Medchem Express HY16935
MedChemExpress Mavatrep is an orally bioavailable TRPV1 antagonist (Ki=6.5 nM), exhibits minimal effect on the enzymatic activity (IC50 > 25 μM) of CYP isoforms 3A4, 1A2, and 2D6.
Mavatrep is an orally bioavailable TRPV1 antagonist (Ki=6.5 nM), exhibits minimal effect on the enzymatic activity (IC50 > 25 μM) of CYP isoforms 3A4, 1A2, and 2D6.
Specifications
Chemical Name or Material Mavatrep
Physical Form Solid
CAS 956274-94-5
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C25H21F3N2O
Synonym JNJ-39439335
Solubility Information DMSO : 16.67 mg/mL (39.46 mM; ultrasonic and warming and heat to 60°C)
SMILES OC(C)(C)C1=CC=CC=C1C2=CC=C3N=C(/C=C/C4=CC=C(C(F)(F)F)C=C4)NC3=C2
Molecular Weight (g/mol) 422.44
Formula Weight 422.44
Percent Purity 98.0%
Grade Research