Learn More
Enclomiphene citrate, MedChemExpress
Product Code. 16431147
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16431147 50 mg 50mg
16457647 5 mg 5mg
16415676 100 mg 100mg
16404898 10 mg 10mg
4 options
Product Code. 16431147
Brand: Medchem Express HY118861A
MedChemExpress Enclomiphene ((E)-Clomiphene) citrate is a potent and orally active non-steroidal estrogen receptor antagonist, with antioestrogenic property. Enclomiphene citrate can be used for the research of ovarian dysfunction, testosterone deficiency, male hypogonadism and type 2 diabetes.
Enclomiphene ((E)-Clomiphene) citrate is a potent and orally active non-steroidal estrogen receptor antagonist, with antioestrogenic property. Enclomiphene citrate can be used for the research of ovarian dysfunction, testosterone deficiency, male hypogonadism and type 2 diabetes.
Specifications
Chemical Name or Material Enclomiphene citrate
CAS 7599-79-3
Color White
Purity Grade Notes Research
Molecular Formula C32H36ClNO8
Quantity 50 mg
Synonym (E)-Clomiphene citratetrans-Clomiphene citrateEnclomifene citrate
Solubility Information DMSO : ≥ 50 mg/mL (83.60 mM)
SMILES Cl/C(C1=CC=CC=C1)=C(C2=CC=C(OCCN(CC)CC)C=C2)\C3=CC=CC=C3.O=C(CC(C(O)=O)(O)CC(O)=O)O.[(E)]
Molecular Weight (g/mol) 598.08
Formula Weight 598.08
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation