Learn More
Necrostatin 2, MedChemExpress
Product Code. 16428717
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16428717
Brand: Medchem Express HY14622
MedChemExpress Necrostatin 2 is a potent necroptosis inhibitor. EC50 for inhibition of necroptosis in FADD-deficient Jurkat T cells treated with TNF-α is 0.05 μM. Necrostatin 2 is also a RIPK1 inhibitor.
Necrostatin 2 is a potent necroptosis inhibitor. EC50 for inhibition of necroptosis in FADD-deficient Jurkat T cells treated with TNF-α is 0.05 μM.
Specifications
Chemical Name or Material Necrostatin 2
Physical Form Solid
CAS 852391-19-6
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C13H12ClN3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (360.09 mM)
SMILES ClC1=C2C(C(C[C@@H]3C(N(C)C(N3)=O)=O)=CN2)=CC=C1
Molecular Weight (g/mol) 277.71
Formula Weight 277.71
Percent Purity 99.53%
Grade Research