Learn More
Org-26576, MedChemExpress
Product Code. 16413308
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16413308 10 mg 10mg
16436027 5 mg 5mg
16424556 100 mg 100mg
16409846 50 mg 50mg
4 options
Product Code. 16413308
Brand: Medchem Express HY101216
MedChemExpress Org-26576 is a AMPA receptor positive allosteric modulator.
Org-26576 is a AMPA receptor positive allosteric modulator.
Specifications
Chemical Name or Material Org-26576
CAS 1026791-61-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H12N2O2
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (122.41 mM; Need ultrasonic)
SMILES O=C1N2[C@](CCC2)([H])COC3=NC=CC=C13
Molecular Weight (g/mol) 204.23
Formula Weight 204.23
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation